Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008791.2 | NP_001008791.1 | 9 - 10 | 2486 | 81 |
NM_001008792.2 | NP_001008792.1 | 8 - 9 | 2465 | 81 | |
NM_001008793.2 | NP_001008793.1 | 8 - 9 | 2453 | 81 | |
NM_028640.2 | NP_082916.1 | 8 - 9 | 2453 | 81 | |
GenBank mRNA | AK122523.1 | - | 8 - 9 | 1584 | 81 |
AY739114.1 | - | 8 - 9 | 2413 | 81 | |
AY739115.1 | - | 9 - 10 | 2446 | 81 | |
AY739116.1 | - | 8 - 9 | 2425 | 81 | |
AY739117.1 | - | 8 - 9 | 2413 | 81 | |
AY739118.1 | - | 5 - 6 | 1062 | 81 | |
AY739119.1 | - | 4 - 5 | 1029 | 81 | |
AY739120.1 | - | 3 - 4 | 816 | 81 | |
AY739121.1 | - | 3 - 4 | 934 | 81 | |
AY739122.1 | - | 4 - 5 | 1266 | 81 | |
BC150757.1 | - | 8 - 9 | 1810 | 81 |