Assay Design



Gene Transcripts

Gene Symbol
Whrn
Entrez Gene ID
73750
Gene Name
whirlin
Gene Aliases
1110035G07Rik, AW122018, AW742671, C430046P22Rik, Dfnb31, wi
Chromosome Location
Chr.4: 63414909 - 63496078 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008791.2 NP_001008791.1 10 - 11 3029 76
NM_001008792.2 NP_001008792.1 9 - 10 3008 76
NM_028640.2 NP_082916.1 9 - 10 2996 76
GenBank mRNA AY739114.1 - 9 - 10 2956 76
AY739115.1 - 10 - 11 2989 76
AY739116.1 - 9 - 10 2968 76
AY739118.1 - 6 - 7 1605 76
AY739119.1 - 5 - 6 1572 76
AY739120.1 - 4 - 5 1359 76
AY739121.1 - 4 - 5 1477 76
AY739122.1 - 5 - 6 1809 76

View Full Product Details