Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008791.2 | NP_001008791.1 | 5 - 6 | 1959 | 75 |
NM_001008793.2 | NP_001008793.1 | 5 - 6 | 1959 | 75 | |
NM_028640.2 | NP_082916.1 | 5 - 6 | 1959 | 75 | |
GenBank mRNA | AK122523.1 | - | 5 - 6 | 1090 | 75 |
AK157955.1 | - | 5 - 6 | 1960 | 75 | |
AY739114.1 | - | 5 - 6 | 1919 | 75 | |
AY739115.1 | - | 5 - 6 | 1919 | 75 | |
AY739117.1 | - | 5 - 6 | 1919 | 75 | |
BC150757.1 | - | 5 - 6 | 1316 | 75 | |
HQ148552.1 | - | 5 - 6 | 1201 | 75 |