Assay Design
Gene Transcripts
- Gene Symbol
- Whrn
- Entrez Gene ID
-
73750
- Gene Name
- whirlin
- Gene Aliases
-
1110035G07Rik, AW122018, AW742671, C430046P22Rik, Dfnb31, wi
- Chromosome Location
-
Chr.4: 63414909 - 63496078 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK021254.1 |
- |
1 - 2 |
154 |
78 |
View Full Product Details