Assay Design



Gene Transcripts

Gene Symbol
Mansc4
Entrez Gene ID
545893
Gene Name
MANSC domain containing 4
Gene Aliases
A430079E08, EG545893, Gm5887
Chromosome Location
Chr.6: 147075062 - 147087032 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034903.3 NP_001030075.1 1 - 2 286 114
GenBank mRNA AK138652.1 - 1 - 2 287 114
BC150953.1 - 1 - 2 261 114

View Full Product Details