Assay Design



Gene Transcripts

Gene Symbol
Elavl2
Entrez Gene ID
15569
Gene Name
ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B)
Gene Aliases
Hub, mel-N1
Chromosome Location
Chr.4: 91250766 - 91400064 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010486.2 NP_034616.1 6 - 7 920 79
GenBank mRNA AY035380.1 - 5 - 6 769 79
U29088.1 - 6 - 7 919 79

View Full Product Details