Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010008.4 | NP_034138.3 | 4 - 5 | 877 | 106 |
GenBank mRNA | AK033614.1 | - | 4 - 5 | 880 | 106 |
AK045421.1 | - | 4 - 5 | 867 | 106 | |
AK084025.1 | - | 4 - 5 | 887 | 106 | |
AK165663.1 | - | 4 - 5 | 722 | 106 | |
BC050832.1 | - | 4 - 5 | 871 | 106 | |
U62295.1 | - | 4 - 5 | 924 | 106 |