Assay Design



Gene Transcripts

Gene Symbol
Aspscr1
Entrez Gene ID
68938
Gene Name
alveolar soft part sarcoma chromosome region, candidate 1 (human)
Gene Aliases
1190006K01Rik, ASPC, ASPCR1, ASPL, ASPS, RCC17, TUG
Chromosome Location
Chr.11: 120672439 - 120709447 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026877.2 NP_081153.1 1 - 2 203 115
GenBank mRNA AK088197.1 - 1 - 2 54 115
AY349133.1 - 1 - 2 120 115
BC019177.1 - 1 - 2 146 115
BC022115.1 - 1 - 2 150 115
BC031153.1 - 1 - 2 135 115

View Full Product Details