Assay Design
Gene Transcripts
- Gene Symbol
- Ccdc68
- Entrez Gene ID
-
381175
- Gene Name
- coiled-coil domain containing 68
- Gene Aliases
-
BC046343
- Chromosome Location
-
Chr.18: 69924858 - 69969484 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_201362.2 |
NP_958750.1 |
9 - 10 |
939 |
129 |
GenBank mRNA |
AK078964.1 |
- |
9 - 10 |
941 |
129 |
View Full Product Details