Assay Design



Gene Transcripts

Gene Symbol
Cd81
Entrez Gene ID
12520
Gene Name
CD81 antigen
Gene Aliases
Tapa-1, Tapa1, Tspan28
Chromosome Location
Chr.7: 143052750 - 143067930 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133655.2 NP_598416.1 1 - 2 321 66
GenBank mRNA AK136370.1 - 1 - 2 285 66
AK166521.1 - 1 - 2 287 66
AK170741.1 - 1 - 2 286 66
AK190176.1 - 1 - 2 202 66
BC011433.1 - 1 - 2 276 66
BY252638.1 - 1 - 2 322 66
X59047.1 - 1 - 2 41 66

View Full Product Details