Assay Design



Gene Transcripts

Gene Symbol
Shf
Entrez Gene ID
435684
Gene Name
Src homology 2 domain containing F
Gene Aliases
-
Chromosome Location
Chr.2: 122348892 - 122369169 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013829.2 NP_001013851.2 5 - 6 908 81
GenBank mRNA BC042839.1 - 5 - 6 796 81
CB056323.1 - 4 - 5 522 81

View Full Product Details