Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001043355.2 | NP_001036820.2 | 1 - 2 | 971 | 63 |
NM_001048167.1 | NP_001041632.1 | 1 - 2 | 1870 | 63 | |
NM_010837.3 | NP_034967.2 | 1 - 2 | 1870 | 63 | |
GenBank mRNA | AK018193.2 | - | 1 - 2 | 1350 | 63 |
AK087413.1 | - | 1 - 2 | 1833 | 63 | |
AK154455.1 | - | 1 - 2 | 943 | 63 | |
BC053039.1 | - | 1 - 2 | 1871 | 63 | |
BY259340.1 | - | 1 - 2 | 193 | 63 | |
Y16032.1 | - | 1 - 2 | 911 | 63 |