Assay Design



Gene Transcripts

Gene Symbol
Ttbk1
Entrez Gene ID
106763
Gene Name
tau tubulin kinase 1
Gene Aliases
AU017937, AW048023, C330008L01Rik
Chromosome Location
Chr.17: 46442448 - 46487675 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162864.1 NP_001156336.1 2 - 3 406 89
GenBank mRNA AK144536.1 - 1 - 2 140 89

View Full Product Details