Assay Design



Gene Transcripts

Gene Symbol
Nt5c1b
Entrez Gene ID
70881
Gene Name
5'-nucleotidase, cytosolic IB
Gene Aliases
4921514H13Rik, AIRP, CN-IB
Chromosome Location
Chr.12: 10369971 - 10390175 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027588.3 NP_081864.2 5 - 6 1045 82
GenBank mRNA AF356186.1 - 5 - 6 964 82
AK014892.1 - 5 - 6 1045 82
BC060976.1 - 5 - 6 983 82

View Full Product Details