Assay Design



Gene Transcripts

Gene Symbol
Ano3
Entrez Gene ID
228432
Gene Name
anoctamin 3
Gene Aliases
AI838058, B230324K02Rik, Tmem16c
Chromosome Location
Chr.2: 110655201 - 110950395 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128103.2 NP_001121575.1 13 - 14 1526 69
GenBank mRNA AK045930.1 - 13 - 14 1526 69

View Full Product Details