Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199113.1 | NP_001186042.1 | 3 - 4 | 267 | 74 |
NM_001199114.1 | NP_001186043.1 | 2 - 3 | 106 | 74 | |
NM_001199115.1 | NP_001186044.1 | 2 - 3 | 212 | 74 | |
NM_001199116.1 | NP_001186045.1 | 2 - 3 | 172 | 74 | |
NM_022880.3 | NP_075018.1 | 1 - 2 | 213 | 74 | |
GenBank mRNA | AF131212.1 | - | 1 - 2 | 214 | 74 |
AF305501.1 | - | 2 - 3 | 177 | 74 | |
AK004756.1 | - | 2 - 3 | 158 | 74 | |
AK050089.1 | - | 2 - 3 | 197 | 74 | |
AK146265.1 | - | 2 - 3 | 107 | 74 | |
AK149483.1 | - | 2 - 3 | 128 | 74 | |
AK170463.1 | - | 2 - 3 | 144 | 74 | |
AK180131.1 | - | 3 - 4 | 204 | 74 | |
AK180168.1 | - | 2 - 3 | 102 | 74 | |
AK191284.1 | - | 3 - 4 | 205 | 74 | |
AK197634.1 | - | 3 - 4 | 204 | 74 | |
AK200370.1 | - | 3 - 4 | 204 | 74 | |
AK205425.1 | - | 3 - 4 | 204 | 74 | |
AK205461.1 | - | 3 - 4 | 204 | 74 | |
AK206588.1 | - | 2 - 3 | 103 | 74 | |
AK218485.1 | - | 1 - 2 | 94 | 74 | |
AK218859.1 | - | 2 - 3 | 103 | 74 | |
BB853265.1 | - | 3 - 4 | 277 | 74 | |
BC004828.1 | - | 3 - 4 | 230 | 74 | |
BC006812.1 | - | 1 - 2 | 144 | 74 | |
BY055687.1 | - | 2 - 3 | 107 | 74 | |
CX219686.1 | - | 3 - 4 | 262 | 74 | |
EU180577.1 | - | 1 - 2 | 82 | 74 |