Assay Design
Gene Transcripts
- Gene Symbol
- B3galt1
- Entrez Gene ID
-
26877
- Gene Name
- UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 1
- Gene Aliases
-
beta3Gal-T1
- Chromosome Location
-
Chr.2: 67565962 - 68122882 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_020283.4 |
NP_064679.2 |
1 - 2 |
51 |
68 |
GenBank mRNA |
AK035215.1 |
- |
1 - 2 |
52 |
68 |
View Full Product Details