Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027959.3 | NP_082235.1 | 9 - 10 | 966 | 80 |
GenBank mRNA | AK005989.1 | - | 9 - 10 | 972 | 80 |
AK076558.1 | - | 9 - 10 | 967 | 80 | |
AK133792.1 | - | 9 - 10 | 974 | 80 | |
AK151008.1 | - | 9 - 10 | 946 | 80 | |
AK152960.1 | - | 9 - 10 | 979 | 80 | |
AK165877.1 | - | 9 - 10 | 945 | 80 | |
AK166111.1 | - | 9 - 10 | 973 | 80 | |
AK167386.1 | - | 9 - 10 | 984 | 80 | |
AK168136.1 | - | 9 - 10 | 973 | 80 | |
AK168279.1 | - | 9 - 10 | 973 | 80 | |
BC006865.1 | - | 9 - 10 | 955 | 80 |