Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024856.2 | NP_001020027.1 | 5 - 6 | 795 | 78 |
NM_001024857.2 | NP_001020028.1 | 4 - 5 | 721 | 78 | |
NM_080788.3 | NP_542966.2 | 5 - 6 | 549 | 78 | |
GenBank mRNA | AB046593.2 | - | 4 - 5 | 473 | 78 |
AK046978.1 | - | 5 - 6 | 768 | 78 | |
AK053820.1 | - | 5 - 6 | 784 | 78 | |
AK077011.1 | - | 5 - 6 | 78 | ||
AK137024.1 | - | 4 - 5 | 710 | 78 | |
AK148269.1 | - | 4 - 5 | 725 | 78 |