Assay Design
Gene Transcripts
- Gene Symbol
- Tmem62
- Entrez Gene ID
-
96957
- Gene Name
- transmembrane protein 62
- Gene Aliases
-
B830009D23Rik, C77355
- Chromosome Location
-
Chr.2: 120976719 - 121007849 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC049167.1 |
- |
2 - 3 |
320 |
68 |
View Full Product Details