Assay Design
Gene Transcripts
- Gene Symbol
- Scn7a
- Entrez Gene ID
-
20272
- Gene Name
- sodium channel, voltage-gated, type VII, alpha
- Gene Aliases
-
1110034K09Rik, AI642000, NaG, Nav2, Nav2.3, Nax, Scn6a
- Chromosome Location
-
Chr.2: 66673424 - 66785091 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_009135.2 |
NP_033161.2 |
21 - 22 |
3686 |
161 |
GenBank mRNA |
L36179.1 |
- |
21 - 22 |
3722 |
161 |
View Full Product Details