Assay Design



Gene Transcripts

Gene Symbol
Pde3b
Entrez Gene ID
18576
Gene Name
phosphodiesterase 3B, cGMP-inhibited
Gene Aliases
9830102A01Rik, AI847709
Chromosome Location
Chr.7: 114414700 - 114537938 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011055.2 NP_035185.2 15 - 16 3383 146
GenBank mRNA AF547435.1 - 15 - 16 3359 149
AJ132271.1 - 15 - 16 3089 149
AK142889.1 - - 269 146
BB663310.1 - 1 - 2 269 146
X95521.1 - 15 - 16 2231 149

View Full Product Details