Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202445.1 | NP_001189374.1 | 11 - 12 | 2475 | 73 |
NM_001202446.1 | NP_001189375.1 | 11 - 12 | 2400 | 73 | |
NM_011162.5 | NP_035292.2 | 11 - 12 | 2183 | 73 | |
GenBank mRNA | AB221603.1 | - | 11 - 12 | 2046 | 73 |
AF003115.1 | - | 11 - 12 | 2042 | 73 | |
AF054611.1 | - | 11 - 12 | 2183 | 73 | |
AF109768.1 | - | 11 - 12 | 2083 | 73 | |
AF109769.1 | - | 11 - 12 | 2146 | 73 | |
AF109770.1 | - | 11 - 12 | 2201 | 73 | |
AF109771.1 | - | 11 - 12 | 2047 | 73 | |
AF332075.1 | - | 11 - 12 | 2046 | 73 | |
AF332076.1 | - | 11 - 12 | 2046 | 73 | |
AK147033.1 | - | 11 - 12 | 2402 | 73 | |
BC072578.1 | - | 11 - 12 | 2367 | 73 |