Assay Design



Gene Transcripts

Gene Symbol
Zcchc24
Entrez Gene ID
71918
Gene Name
zinc finger, CCHC domain containing 24
Gene Aliases
2310047A01Rik
Chromosome Location
Chr.14: 25711640 - 25768856 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101433.1 NP_001094903.1 1 - 2 245 88
GenBank mRNA BC147263.1 - 1 - 2 335 88
BC147264.1 - 1 - 2 335 88

View Full Product Details