Assay Design
Gene Transcripts
- Gene Symbol
- Cnpy3
- Entrez Gene ID
-
72029
- Gene Name
- canopy 3 homolog (zebrafish)
- Gene Aliases
-
1600025D17Rik, 2410050O22Rik, AI413153, CAG4A, ERDA5, PRAT4A, Tnrc5
- Chromosome Location
-
Chr.17: 46735713 - 46752293 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK082340.1 |
- |
1 - 2 |
223 |
63 |
View Full Product Details