Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025878.1 | NP_080154.1 | 3 - 4 | 289 | 58 |
GenBank mRNA | AB049954.1 | - | 3 - 4 | 313 | 58 |
AK010250.1 | - | 3 - 4 | 296 | 58 | |
AK088540.1 | - | 3 - 4 | 285 | 58 | |
AK166597.1 | - | 3 - 4 | 310 | 58 | |
AK167067.1 | - | 3 - 4 | 290 | 58 | |
BC021752.1 | - | 3 - 4 | 289 | 58 |