Assay Design



Gene Transcripts

Gene Symbol
Mdc1
Entrez Gene ID
240087
Gene Name
mediator of DNA damage checkpoint 1
Gene Aliases
6820401C03, AA413496, Nfbd1, mKIAA0170
Chromosome Location
Chr.17: 35841490 - 35859670 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010833.2 NP_001010833.2 15 - 16 5151 65
GenBank mRNA AK129074.1 - 9 - 10 2887 65
BC085140.1 - 10 - 11 3020 65
BC094363.1 - 11 - 12 3854 65

View Full Product Details