Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001082552.2 | NP_001076021.1 | 6 - 7 | 71 | |
NM_009277.4 | NP_033303.3 | 7 - 8 | 71 | ||
GenBank mRNA | AK141294.1 | - | 7 - 8 | 71 | |
AK150484.1 | - | 6 - 7 | 71 | ||
AK152619.1 | - | 6 - 7 | 71 | ||
AK171489.1 | - | 7 - 8 | 71 | ||
BC010580.1 | - | 7 - 8 | 991 | 71 | |
CT010336.1 | - | 5 - 6 | 876 | 71 | |
L27990.1 | - | 7 - 8 | 1016 | 71 |