Assay Design



Gene Transcripts

Gene Symbol
Art1
Entrez Gene ID
11870
Gene Name
ADP-ribosyltransferase 1
Gene Aliases
ADPRT, ARTC1, Yac-1
Chromosome Location
Chr.7: 102101706 - 102111758 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009710.4 NP_033840.2 1 - 2 43 66
GenBank mRNA AK003107.1 - 1 - 2 44 66
AK009284.1 - 1 - 2 44 66
AK009902.1 - - 44 66

View Full Product Details