Assay Design
Gene Transcripts
- Gene Symbol
- Art1
- Entrez Gene ID
-
11870
- Gene Name
- ADP-ribosyltransferase 1
- Gene Aliases
-
ADPRT, ARTC1, Yac-1
- Chromosome Location
-
Chr.7: 102101706 - 102111758 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK009284.1 |
- |
5 - 6 |
711 |
98 |
View Full Product Details