Assay Design



Gene Transcripts

Gene Symbol
Tpp1
Entrez Gene ID
12751
Gene Name
tripeptidyl peptidase I
Gene Aliases
Cln2, TPP-I
Chromosome Location
Chr.7: 105744847 - 105752282 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009906.5 NP_034036.1 7 - 8 905 86
GenBank mRNA AF111172.1 - 7 - 8 913 86
AJ011912.1 - 7 - 8 899 86
AK002418.1 - 7 - 8 924 86
AK048279.1 - 7 - 8 906 86
AK083832.1 - 6 - 7 2170 86
AK155021.1 - 7 - 8 924 86
AK165669.1 - 7 - 8 930 86
AK166648.1 - 5 - 6 698 85
AK169924.1 - 7 - 8 912 86
AK170781.1 - 7 - 8 901 86
BC024820.1 - 7 - 8 907 86

View Full Product Details