Assay Design



Gene Transcripts

Gene Symbol
Atp2a1
Entrez Gene ID
11937
Gene Name
ATPase, Ca++ transporting, cardiac muscle, fast twitch 1
Gene Aliases
SERCA1
Chromosome Location
Chr.7: 126445860 - 126463103 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007504.2 NP_031530.2 4 - 5 477 61
GenBank mRNA AY081946.1 - 4 - 5 508 61
BC036292.1 - 4 - 5 477 61

View Full Product Details