Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009383.2 | NP_033409.1 | 11 - 12 | 1410 | 58 |
GenBank mRNA | AK012085.1 | - | 11 - 12 | 1054 | 58 |
AK054473.1 | - | 4 - 5 | 806 | 58 | |
AK079363.1 | - | 1 - 2 | 57 | 58 | |
AK132677.1 | - | 12 - 13 | 1578 | 58 | |
BC010496.1 | - | 12 - 13 | 1144 | 58 | |
BC068194.1 | - | 7 - 8 | 686 | 58 | |
U55861.1 | - | 11 - 12 | 1186 | 58 |