Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009760.4 | NP_033890.1 | 5 - 6 | 631 | 86 |
GenBank mRNA | AF041054.1 | - | 5 - 6 | 601 | 86 |
AK014223.1 | - | 5 - 6 | 632 | 86 | |
AK075943.1 | - | 5 - 6 | 632 | 86 | |
AK152610.1 | - | 5 - 6 | 627 | 86 | |
AK170865.1 | - | 5 - 6 | 595 | 86 | |
BC033278.1 | - | 5 - 6 | 539 | 86 | |
BC046603.1 | - | 5 - 6 | 621 | 86 |