Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009739.3 | NP_033869.1 | 11 - 12 | 1364 | 78 |
GenBank mRNA | AF043070.1 | - | 10 - 11 | 1205 | 78 |
AK075602.1 | - | 11 - 12 | 1290 | 78 | |
AK145479.1 | - | 11 - 12 | 1335 | 78 | |
AK150607.1 | - | 11 - 12 | 1356 | 78 | |
AK150723.1 | - | 11 - 12 | 1356 | 78 | |
AK167519.1 | - | 11 - 12 | 1356 | 78 | |
BC004077.1 | - | 8 - 9 | 792 | 78 | |
BC046595.1 | - | 11 - 12 | 1334 | 78 |