Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177576.1 | NP_001171047.1 | 7 - 8 | 810 | 96 |
NM_026646.3 | NP_080922.1 | 7 - 8 | 1052 | 96 | |
GenBank mRNA | AK010193.1 | - | 7 - 8 | 798 | 96 |
AK018760.1 | - | 7 - 8 | 835 | 96 | |
AK046874.1 | - | 7 - 8 | 797 | 96 | |
AK048389.1 | - | 7 - 8 | 781 | 96 | |
AK049313.1 | - | 7 - 8 | 781 | 96 | |
AK155276.1 | - | 7 - 8 | 804 | 96 | |
AK162400.1 | - | 7 - 8 | 763 | 96 | |
BC037949.1 | - | 7 - 8 | 809 | 96 | |
BC050887.1 | - | 6 - 7 | 1528 | 96 |