Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026992.3 | NP_081268.1 | 2 - 3 | 606 | 142 |
GenBank mRNA | AK006521.1 | - | 2 - 3 | 607 | 142 |
AY028460.1 | - | 2 - 3 | 288 | 142 | |
BC030072.1 | - | 2 - 3 | 314 | 142 | |
BC091774.1 | - | 2 - 3 | 172 | 142 |