Assay Design
Gene Transcripts
- Gene Symbol
- Nadsyn1
- Entrez Gene ID
-
78914
- Gene Name
- NAD synthetase 1
- Gene Aliases
-
9130012B15Rik
- Chromosome Location
-
Chr.7: 143795587 - 143822864 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC018248.1 |
- |
6 - 7 |
536 |
86 |
View Full Product Details