Assay Design



Gene Transcripts

Gene Symbol
Fgf15
Entrez Gene ID
14170
Gene Name
fibroblast growth factor 15
Gene Aliases
FGF19
Chromosome Location
Chr.7: 144896532 - 144900953 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008003.2 NP_032029.1 1 - 2 398 88
GenBank mRNA AF007268.1 - 1 - 2 398 88
AK017829.1 - 1 - 2 400 88
BC021328.1 - 1 - 2 390 88

View Full Product Details