Assay Design



Gene Transcripts

Gene Symbol
Slc24a3
Entrez Gene ID
94249
Gene Name
solute carrier family 24 (sodium/potassium/calcium exchanger), member 3
Gene Aliases
NCKX3
Chromosome Location
Chr.2: 145167712 - 145642166 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053195.2 NP_444425.1 3 - 4 253 89
GenBank mRNA AF314821.1 - 3 - 4 293 89
BC017615.1 - 3 - 4 253 89

View Full Product Details