Assay Design



Gene Transcripts

Gene Symbol
Nt5c1b
Entrez Gene ID
70881
Gene Name
5'-nucleotidase, cytosolic IB
Gene Aliases
4921514H13Rik, AIRP, CN-IB
Chromosome Location
Chr.12: 10369971 - 10390175 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027588.3 NP_081864.2 4 - 5 880 78
GenBank mRNA AJ295253.1 - 4 - 5 807 78
AK014892.1 - 4 - 5 880 78

View Full Product Details