Assay Design



Gene Transcripts

Gene Symbol
Pdcd2
Entrez Gene ID
18567
Gene Name
programmed cell death 2
Gene Aliases
AI528543, Pcd2, RP-8
Chromosome Location
Chr.17: 15521575 - 15527301 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008799.2 NP_032825.2 3 - 4 684 87
GenBank mRNA AK045985.1 - 3 - 4 685 87
DQ906042.1 - - 664 87
U10903.1 - 3 - 4 664 87

View Full Product Details