Assay Design
Gene Transcripts
- Gene Symbol
- Mff
- Entrez Gene ID
-
75734
- Gene Name
- mitochondrial fission factor
- Gene Aliases
-
5230400G24Rik, AI314724
- Chromosome Location
-
Chr.1: 82724895 - 82752390 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_029409.2 |
NP_083685.2 |
5 - 6 |
759 |
118 |
GenBank mRNA |
BC059229.1 |
- |
5 - 6 |
759 |
118 |
View Full Product Details