Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289451.1 | NP_001276380.1 | 4 - 5 | 624 | 66 |
NM_001289457.1 | NP_001276386.1 | 4 - 5 | 624 | 66 | |
NM_027763.2 | NP_082039.1 | 4 - 5 | 639 | 66 | |
NR_110338.1 | 4 - 5 | 395 | 66 | ||
GenBank mRNA | AK017256.1 | - | 4 - 5 | 625 | 66 |
AY078502.1 | - | 4 - 5 | 639 | 66 | |
AY286071.1 | - | 4 - 5 | 624 | 66 | |
BC138399.1 | - | 4 - 5 | 623 | 66 | |
BC145263.1 | - | 4 - 5 | 623 | 66 | |
DQ087181.1 | - | 4 - 5 | 392 | 66 | |
DQ087182.1 | - | 3 - 4 | 773 | 66 | |
DQ087183.1 | - | 4 - 5 | 621 | 66 |