Assay Design



Gene Transcripts

Gene Symbol
Ipo7
Entrez Gene ID
233726
Gene Name
importin 7
Gene Aliases
A330055O14Rik, C330016G14, Imp7, Ranbp7
Chromosome Location
Chr.7: 110018227 - 110056610 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181517.3 NP_852658.2 14 - 15 1586 81
GenBank mRNA AK039534.1 - 14 - 15 1738 81
AK079903.1 - 13 - 14 3096 81
AY316153.1 - 14 - 15 1586 81
BC053524.1 - 11 - 12 1269 81
BC064825.1 - 5 - 6 542 81

View Full Product Details