Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026115.4 | NP_080391.2 | 2 - 3 | 143 | 120 |
GenBank mRNA | AK010715.1 | - | 2 - 3 | 152 | 120 |
AK014330.1 | - | 2 - 3 | 155 | 120 | |
AK018270.1 | - | 2 - 3 | 138 | 120 | |
AK041700.1 | - | 2 - 3 | 147 | 120 | |
AK137648.1 | - | 2 - 3 | 161 | 120 | |
AK169315.1 | - | 2 - 3 | 146 | 120 | |
AK182724.1 | - | 1 - 2 | 100 | 120 | |
BC055460.1 | - | 1 - 2 | 110 | 120 |