Assay Design



Gene Transcripts

Gene Symbol
Zc3h6
Entrez Gene ID
78751
Gene Name
zinc finger CCCH type containing 6
Gene Aliases
4631426G04Rik, 4833425H18Rik, AU024389, AW060453, Zc3hdc6, mKIAA2035
Chromosome Location
Chr.2: 128967402 - 129018563 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178404.3 NP_848491.2 9 - 10 1737 164
GenBank mRNA AK019473.1 - 2 - 3 282 164
AK039171.1 - 9 - 10 1732 164
BC043311.1 - 5 - 6 693 164

View Full Product Details