Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175467.3 | NP_780676.1 | 12 - 13 | 1746 | 78 |
GenBank mRNA | AK048374.1 | - | 11 - 12 | 1607 | 78 |
AK054240.1 | - | 11 - 12 | 1833 | 78 | |
AK078679.1 | - | 11 - 12 | 1810 | 78 | |
AK078686.1 | - | 12 - 13 | 1747 | 78 | |
BC094496.1 | - | 11 - 12 | 1591 | 78 |