Assay Design



Gene Transcripts

Gene Symbol
Slc5a7
Entrez Gene ID
63993
Gene Name
solute carrier family 5 (choline transporter), member 7
Gene Aliases
CHT1
Chromosome Location
Chr.17: 54273590 - 54299041 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022025.4 NP_071308.2 7 - 8 1408 82
GenBank mRNA AF276872.2 - 6 - 7 899 82
AJ401467.1 - 6 - 7 899 82
AK034415.1 - 7 - 8 1292 82
AK053063.1 - 7 - 8 1410 82
BC065089.1 - 6 - 7 925 82

View Full Product Details