Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145526.2 | NP_663501.2 | 11 - 12 | 1611 | 105 |
GenBank mRNA | AK140283.1 | - | 11 - 12 | 2879 | 105 |
AK141847.1 | - | 11 - 12 | 1276 | 105 | |
BC023089.1 | - | 11 - 12 | 1264 | 105 |